Beryllium ≥99.9% (metals basis), ultrathin foil, Thickness: 1 µm

Toimittaja: Thermo Fisher Scientific
Vaara

40709.FF 40709.FI
40709.FFEA 0 EUR
40709.FF 40709.FI
Beryllium ≥99.9% (metals basis), ultrathin foil, Thickness: 1 µm
Beryllium

Purity: 99.9% CAS No.: 7440-41-7 Physical Form: On temporary copper support Odour: Odourless MDL No.: MFCD00134032


Alloy source, X-ray tube manufacture, neutron source

Kaava: Be
MW: 9.01 g/mol
Kiehumispiste: 2970
Sulamispiste: 1278
Tiheys: 1.85 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134032
CAS-numero: 7440-41-7
EINECS: 231-150-7
Merck Index: 12,01201

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR