Nikkeli ≥99.98% (metals basis), lanka, hard, Ø 0.01 mm (0.0004 in)

Toimittaja: Thermo Fisher Scientific
Vaara

40673.G1
40673.G1EA 774 EUR
40673.G1
Nikkeli ≥99.98% (metals basis), lanka, hard, Ø 0.01 mm (0.0004 in)
Nikkeli

Purity: 99.98% CAS No.: 7440-02-0 Physical Form: ≈0.0007g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011137


High temperature and corrosion resistant materials

Kaava: Ni
Kiehumispiste: 3007 °C (1013 hPa)
Sulamispiste: 1453 °C
Tiheys: 8,9 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011137
CAS-numero: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR