Vismutti(III)nitraatti, emäksinen (≥79% Bi₂O₃)

Toimittaja: Thermo Fisher Scientific
Vaara

40476.14 40476.22 40476.36
40476.14EA 94 EUR
40476.14 40476.22 40476.36
Vismutti(III)nitraatti, emäksinen (≥79% Bi₂O₃)
Vismutti(III)nitraatti, emäksinen

CAS No.: 1304-85-4 Molecular Formula: Bi5O(OH)9(NO3)4 (approximate) Formula Weight: 1461.98 Physical Form: Powder Density: 6.04 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 260° dec. Solubility: Practically insoluble in water and alcohol. Soluble in dilute HNO3 and HCl Merck Reference: 14,1284 UN No.: 1477 MDL No.: MFCD00064844


Ceramic glazes and enamel fluxes

Kaava: Bi₅O(OH)₉(NO₃)₄
Sulamispiste: 260 °C (haj. lämpötila)
Tiheys: 6,04
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064844
CAS-numero: 1304-85-4
EINECS: 215-136-8
UN: 1477
ADR: 5.1,II
Merck Index: 14,1284

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR