Nikkelidibromiditrihydraatii ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

40242.18 40242.30
40242.18EA 285 EUR
40242.18 40242.30
Nikkelidibromiditrihydraatii ≥98%
Nikkelidibromiditrihydraatii

Purity: 98% CAS No.: 7789-49-3 Molecular Formula: NiBr2.3H2O Formula Weight: 272.58 (218.53anhy) Physical Form: Crystalline Granular Sensitivity: Hygroscopic Solubility: Soluble in water, alcohol Merck Reference: 14,6502 UN No.: 3077 MDL No.: MFCD00011141

Kaava: Br₂Ni·3H₂O
Sulamispiste: 300 °C (-3H₂O)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011141
CAS-numero: 7789-49-3
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Merck Index: 14,6502

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR