Selenium tetrabromide 99

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Selenium(IV) bromide

40240.14 40240.06
40240.14EA 532 EUR
40240.14 40240.06
Selenium tetrabromide 99
Selenium tetrabromide

Purity: 99+% CAS No.: 7789-65-3 Molecular Formula: SeBr4 Formula Weight: 398.60 Physical Form: Powder Sensitivity: Moisture Sensitive Solubility: Decomposes in water. Soluble in carbon disulfide, chloroform, ethyl bromide Merck Reference: 14,8437 UN No.: 2923 MDL No.: MFCD00049766

Kaava: Br₄Se
MW: 398.58 g/mol
Sulamispiste: 70…80 °C (haj. lämpötila)
Tiheys: 4.029 g/cm³ (–78 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049766
CAS-numero: 7789-65-3
EINECS: 232-181-9
UN: 2923
ADR: 8 II
Merck Index: 12,08581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR