Palladium 99,98 Metals basis, wire, Ø 1.0 mm

Toimittaja: Thermo Fisher Scientific

40212.BS 40212.BW 40212.CB
40212.BSEA 741 EUR
40212.BS 40212.BW 40212.CB
Palladium 99,98 Metals basis, wire, Ø 1.0 mm
Palladium

Purity: 99.98+% CAS No.: 7440-05-3 Physical Form: ≈9.44g/m Odour: Odourless MDL No.: MFCD00011167

Kaava: Pd
MW: 106.420 g/mol
Tiheys: 12.02 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011167
CAS-numero: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR