Tetrakloorisilaani ≥99.998% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

40148.ACEA 134 EUR
40148.AC 40148.AE
Tetrakloorisilaani ≥99.998% (metals basis)
Tetrakloorisilaani

Purity: 99.998% CAS No.: 10026-04-7 Molecular Formula: Cl4Si Formula Weight: 169.90 Physical Form: Liquid Density: 1.483 Appearance: Colourless Odour: Acrid Melting Point: -70° Boiling Point: 57.6° Merck Reference: 14,8500 UN No.: 1818 MDL No.: MFCD00011229

Kaava: Cl₄Si
Kiehumispiste: 57,6 °C
Sulamispiste: –70 °C
Tiheys: 1,483
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011229
CAS-numero: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 14,8500

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR