Natriummetabisulfiitti ≥97% ACS

Toimittaja: Thermo Fisher Scientific
Vaara

40115.36 40115.A4
40115.36EA 113 EUR
40115.36 40115.A4
Natriummetabisulfiitti ≥97% ACS
Natriummetabisulfiitti

Purity: 97% min CAS No.: 7681-57-4 Molecular Formula: Na2O5S2 Formula Weight: 190.10 Physical Form: Crystalline powder Density: 1.480 Appearance: White to pale yellow Odour: Like rotten eggs pH: 3.5-5.0 Melting Point: 300° Merck Reference: 14,8638 MDL No.: MFCD00167602

Kaava: Na₂S₂O₅
Sulamispiste: >300 °C
Tiheys: 1,480
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00167602
CAS-numero: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Index: 14,8638
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR