Nickel(II) carbonate (basic) hydrate (≥40% Ni) ≥99.5% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

40106.30 40106.A1
40106.30EA 146 EUR
40106.30 40106.A1
Nickel(II) carbonate (basic) hydrate (≥40% Ni) ≥99.5% (metals basis)
Nickel(II) carbonate (basic) hydrate

Purity: 99.5% CAS No.: 155775-31-8 Molecular Formula: CNiO3.2H2NiO2.xH2O Formula Weight: 304.18(anhy) Physical Form: Powder Appearance: Green Odour: Odourless UN No.: 3077 MDL No.: MFCD00016262


Nickel plating pH adjuster, catalysts, ceramic Colours and glazes

Kaava: CNiO₃·2H₂NiO₂·xH₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016262
CAS-numero: 155775-31-8
EINECS: 240-408-8
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR