Anti-CD3E Rat Monoclonal Antibody [clone: 17A2]
Toimittaja: Tonbo Biosciences
40-0032-U500 40-0032-U100
40-0032-U500EA 142 EUR
40-0032-U500 40-0032-U100
Anti-CD3E Rat Monoclonal Antibody [clone: 17A2]
Vasta-aineet
The 17A2 antibody reacts with the mouse CD3 complex, comprised of CD3 epsilon, CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines orMore Product Information