Trifluorietikkahappo 0,05% v/v acetonitrile, LiChrosolv® HPLC, Supelco®

Toimittaja: Merck
Vaara

Synonyms: TFA

MQ100

4.80672.2500
4.80672.2500EA 349 EUR
4.80672.2500
Trifluorietikkahappo 0,05% v/v acetonitrile, LiChrosolv® HPLC, Supelco®
Trifluorietikkahappo
Kaava: C₂HF₃O₂
MW: 114.02 g/mol
Kiehumispiste: 71.8
Sulamispiste: -15
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004169
CAS-numero: 76-05-1
EINECS: 200-929-3
UN: 1648
ADR: 3 II
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR