Trifluorietikkahappo 0,1% (v/v) in aqueous solution, LiChrosolv®, hypergrade for HPLC, Supelco®

Toimittaja: Merck

Synonyms: TFA

M-Clarity™ quality level = MQ100

4.80112.2500 4.80112.9030 4.80112.4000
4.80112.2500EA 97.5 EUR
4.80112.2500 4.80112.9030 4.80112.4000
Trifluorietikkahappo 0,1% (v/v) in aqueous solution, LiChrosolv®, hypergrade for HPLC, Supelco®
Trifluorietikkahappo

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to perfectly meet LC-MS quality requirements.

Kaava: C₂HF₃O₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004169
CAS-numero: 76-05-1
EINECS: 200-929-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR