1-Butanoli, extra dry 99+%, AcroSeal®

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

398960010 398961000
398960010.EA 190 EUR
398960010. 398961000.
1-Butanoli, extra dry 99+%, AcroSeal®
1-Butanoli
Kaava: H₃C(CH₂)₂CH₂OH
Kiehumispiste: 117,6 °C (760 mmHg)
Sulamispiste: –89 °C
Tiheys: 0,81 g/cm³
Leimahduspiste: 35 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002964
CAS-numero: 71-36-3
UN: 1120
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR