Kupari ≥99.99% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)

Toimittaja: Thermo Fisher Scientific

39674.18 39674.30
39674.18EA 191 EUR
39674.18 39674.30
Kupari ≥99.99% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Kupari

Purity: 99.99% CAS No.: 7440-50-8 Physical Form: ≈1.8g/slug Odour: Odourless MDL No.: MFCD00010965

Kaava: Cu
Kiehumispiste: 2595 °C (1013 hPa)
Sulamispiste: 1083 °C
Tiheys: 8,94 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010965
CAS-numero: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR