Thallium(I) hexafluorophosphate 97

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Thallium(I) hexafluorophosphate(V)

39627.06 39627.14
39627.06EA 636 EUR
39627.06 39627.14
Thallium(I) hexafluorophosphate 97
Thallium(I) hexafluorophosphate

Purity: 97% min CAS No.: 60969-19-9 Molecular Formula: TlPF6 Formula Weight: 349.34 Physical Form: Crystalline UN No.: 1707 MDL No.: MFCD00049807

Kaava: F₆PTl
MW: 349.344 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049807
CAS-numero: 60969-19-9
UN: 1707
ADR: 6.1 II

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR