Tin(II) trifluoromethanesulphonate ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

39603.03 39603.06
39603.03EA 118 EUR
39603.03 39603.06
Tin(II) trifluoromethanesulphonate ≥97%
Tin(II) trifluoromethanesulphonate

Purity: 97% CAS No.: 62086-04-8 Molecular Formula: Sn(CF3SO3)2 Formula Weight: 416.82 Physical Form: Powder Sensitivity: Moisture Sensitive UN No.: 3288 MDL No.: MFCD00191251

Kaava: C₂F₆O₆S₂Sn
Sulamispiste: Dec.
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00191251
CAS-numero: 62086-04-8
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR