Tetrachlorobis(tetrahydrofuran)zirconium(IV)

Toimittaja: Thermo Fisher Scientific
Vaara

39550.04 39550.09
39550.04EA 110 EUR
39550.04 39550.09
Tetrachlorobis(tetrahydrofuran)zirconium(IV)
Tetrachlorobis(tetrahydrofuran)zirconium(IV)

CAS No.: 21959-01-3 Molecular Formula: C8H16Cl4O2Zr Formula Weight: 377.25 Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: White to off-white Melting Point: 175-177° UN No.: 2925 MDL No.: MFCD00145367

Kaava: C₄H₈Cl₄OZr
Sulamispiste: 175…177 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00145367
CAS-numero: 21959-01-3
UN: 2925
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR