Iridium(IV) bromide ≥99.99% (metals basis) Ir ≥ 37.1%, Premion®

Toimittaja: Thermo Fisher Scientific
Varoitus

39493.02 39493.04
39493.02EA 359 EUR
39493.02 39493.04
Iridium(IV) bromide ≥99.99% (metals basis) Ir ≥ 37.1%, Premion®
Iridium(IV) bromide

Purity: 99.99% CAS No.: 7789-64-2 Molecular Formula: IrBr4 Formula Weight: 511.84 Physical Form: Crystalline Odour: Acrid MDL No.: MFCD00049467

Kaava: Br₄Ir
Sulamispiste: Dec.
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049467
CAS-numero: 7789-64-2
EINECS: 232-180-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR