Iron yttrium oxide 99,9

Toimittaja: Thermo Fisher Scientific
Varoitus

39488.22
39488.22EA 866 EUR
39488.22
Iron yttrium oxide 99,9
Iron yttrium oxide

Purity: 99.9% CAS No.: 12063-56-8 Molecular Formula: Y3Fe5O12 Formula Weight: 737.94 Physical Form: Powder Odour: Odourless MDL No.: MFCD00210618

Kaava: Y₃Fe₅O₁₂
MW: 737.94 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00210618
CAS-numero: 12063-56-8
EINECS: 235-053-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR