magnesium pyrophosphate

Toimittaja: Thermo Fisher Scientific
Varoitus

39317.18 39317.30 39317.A1
39317.18EA 140 EUR
39317.18 39317.30 39317.A1
magnesium pyrophosphate
magnesium pyrophosphate

CAS No.: 13446-24-7 Molecular Formula: Mg2P2O7 Formula Weight: 222.57 Physical Form: Powder Appearance: White MDL No.: MFCD00049482

Kaava: Mg₂O₇P₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049482
CAS-numero: 13446-24-7
EINECS: 236-595-0
Merck Index: 14,5684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR