VWR®, Cell culture media, RPMI 1640
Toimittaja: VWR Chemicals
392-0425EA 10.8 EUR
392-0425 392-0426 392-0427 392-0428 392-0429 392-0430
VWR®, Cell culture media, RPMI 1640
Soluviljelyyn liuokset
RPMI are general purpose enriched media. Originally formulated for suspension cultures and monolayer cultures of human leukemic cells.
  • RPMI media are 'multipurpose' enriched media, providing extensive applications for a broad spectrum of mammalian and hybridoma cells including:
  • Human myeloma
  • Mouse hybridoma
  • Human leukocytes
  • B and T lymphocytes