Trypsinizing flasks, WHEATON®
Toimittaja: DWK Life Sciences
355803 355804 355807 355754 355753 355752 355755 355757
WHEA355803EA 328 EUR
WHEA355803 WHEA355804 WHEA355807 WHEA355754 734-3116 392-0369 WHEA355755 WHEA355757
Trypsinizing flasks, WHEATON®
Kolvit ja erlenmeyerit Trypsinaatiopullot
Borosilicate glass, clear.
  • With pour out and caps
  • Fluted design improves mixing
  • Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.