Tin(IV) 2-ethylhexanoate diisopropoxide ≥98% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

39169.03 39169.06
39169.03EA 86.6 EUR
39169.03 39169.06
Tin(IV) 2-ethylhexanoate diisopropoxide ≥98% (metals basis)
Tin(IV) 2-ethylhexanoate diisopropoxide

Purity: 98% CAS No.: 288374-32-3 Molecular Formula: C22H44O6Sn Formula Weight: 523.08 Physical Form: Liquid Sensitivity: Moisture Sensitive MDL No.: MFCD00210604

Kaava: C₂₂H₄₄O₆Sn
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00210604
CAS-numero: 288374-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR