Multi-element QC-19 check standard solution, Specpure®

Toimittaja: Thermo Fisher Scientific

39146.AE 39146.AP
39146.AEEA 1030 EUR
39146.AE 39146.AP
Multi-element QC-19 check standard solution, Specpure®
Standardiaineet ja vertailumateriaalit Cationic Standards Multi-Element Standards

Molecular Formula: Matrix: 5% HNO3/tr. F-/tr. tartaric acid
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00210655

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR