HEPES free acid, ULTROL®, Millipore®

Toimittaja: Merck Millipore Calbiochem

391338-1 391338-100 391338-25 391338-5 391338-500
391338-1EA 875 EUR
391338-1 391338-100 391338-25 391338-5 391338-500
HEPES free acid, ULTROL®, Millipore®
HEPES
Kaava: C₈H₁₈N₂O₄S
Sulamispiste: 210…215 °C
Tiheys: 1,448 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006158
CAS-numero: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR