PCR tubes for MicroRack tube system
Toimittaja: Greiner Bio-One
102201_1000 102270_1000 102261_1000 365261_120
391-3721EA 94.4 EUR
391-3721 391-3722 391-3723 391-3725
PCR tubes for MicroRack tube system
Putket Koeputket
PP tubes designed to work with the MicroRack tube storage system.
  • Polypropylene tubes have particularly good thermal, chemical and mechanical stability
  • Ideally suited for the storage of chemical and biological samples