Culture tubes with screw cap, C-TUBE®, WHEATON®
Toimittaja: DWK Life Sciences
358646 358650 358647 358606 358607
391-0197EA 623 EUR
391-0197 391-0198 WHEA358647 WHEA358606 WHEA358607
Culture tubes with screw cap, C-TUBE®, WHEATON®
Putket Viljelyputket
Manufactured from 51 expansion borosilicate glass that conforms to ASTM Type I, Class B and USP Type I requirements.
  • For use as slant or shake cultures
  • Autoclavable
  • Very good chemical resistance
The tubes can be reused many times. The caps have a white 14B styrene-butadiene rubber liner or a PTFE faced 14B styrene-butadiene rubber liner.