Constant Climate Chambers, KBF Series
Toimittaja: Binder
9020-0478 9020-0479 9020-0480 9020-0481 9020-0482 9020-0483
390-1753EA 16960 EUR
390-1753 390-1754 390-1755 390-1756 390-1757 390-1758
Constant Climate Chambers, KBF Series
Testikammiot Olosuhdekaapit
The KBF series is a proven specialist for reliable ICH stability tests in the pharmaceutical industry: the KBF masters all long-term tests, accelerated tests and stress tests. It fulfills all important ICH requirements, from precision to programming and documentation. With flexible water supply options for all installation locations. The units have a stainless-steel inner chamber and proven APT.line™ preheating chamber technology, feature a temperature range of 0 to +70 °C, and humidity range of 10 to 80% RH, the KBF consumes up to 40 % less energy than its predecessor. With optionalMore Product Information