Heratherm™ CO₂ Incubators
Toimittaja: Thermo Fisher Scientific
ECS750 ECG750
390-1716EA 25410 EUR
390-1716 390-1717
Heratherm™ CO₂ Incubators
Inkubaattorit CO₂-inkubaattorit
Heratherm™ CO₂ incubators offer exceptional environmental control in a hassle-free chamber that is easy to program and use.
  • Reduce your environmental footprint while achieving outstanding results with Heratherm CO₂ incubators decreased water and energy consumption and Peltier technology
  • Water-efficient humidification system with integrated water tank and optional clean water connection
  • Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors
  • Different models available offering various