Cooled incubators with environmentally friendly thermoelectric cooling, KB ECO series
Toimittaja: Binder
9020-0423 9020-0424 9020-0425 9020-0465
390-1641EA 9000 EUR
390-1641 390-1642 390-1643 390-1695
Cooled incubators with environmentally friendly thermoelectric cooling, KB ECO series
Inkubaattorit Jäähdytyskaapit
Thanks to thermoelectric cooling technology with heat dissipation, KB ECO cooled incubators are one of the most energy efficient cooling incubators on the market. The thermoelectric cooling technology means the KB ECO series is also very quiet during operation.
  • Safe: Unique condensate and dry-out protection for all operating conditions
  • Reliable: Fail-safe operation due to intelligent thermoelectric tempering
  • Smart: Temperatures programmable for intermitted incubation and refrigeration
  • Economical: Minimum energy consumption thanks to optimised