Cooled incubators, HettCube Range
Toimittaja: Hettich
62005 64005 66005
390-0971EA 8600 EUR
390-0971 390-0973 390-0975
Cooled incubators, HettCube Range
Inkubaattorit Jäähdytyskaapit
The HettCube range of cooled incubators provide functionality and customised features. The inner chamber is made from acid-proof stainless steel (DIN 1.4301). Units only heat and cool as necessary, ensuring lower power consumption, operating costs and environmental impact. At an operating temperature of 37 °C the units consume less than 0,05 kWh. The fan is located outside the usable space, so there is a low air flow inside the chamber and a low noise level.
  • Microprocessor control with program functions, temperatures can be set with an accuracy of 0,1 °C