Incubators with natural convection (BD) or forced convection (BF), Avantgarde.Line
Toimittaja: Binder
9010-0313 9010-0323 9010-0325 9010-0329 9010-0315 9010-0319 9010-0331 9010-0321
390-0921EA 2410 EUR
390-0921 390-0918 390-0919 390-0920 390-0922 390-0923 BIND9010-0331 BIND9010-0321
Incubators with natural convection (BD) or forced convection (BF), Avantgarde.Line
Inkubaattorit Perusinkubaattorit
Avantgarde. Line incubators ensure stable and reliable incubation of microbiological cultures and guarantee reproducible results for routine tests in the laboratory – even high batch throughputs and with long-term operation. Avantgarde. Line BD models have natural convection ensuring rapid temperature equalisation after the door is opened. The BF models have forced convection via a fan. The APT.line™ pre-heating chamber technology gives homogeneous temperature distribution, excellent temperature uniformity and low fluctuation. Units up to 115 litres can be stacked on top of each other toMore Product Information