Naphthol AS-BI phosphate solution 12,5 mg/mL, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY

Synonyms: 7-Bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, Naphthol AS-BI Phosphoric acid solution, Naphthol AS-BI-Phosphate, 6-Bromo-3-((2-methoxyphenyl)carbamoyl)naphthalen-2-yl dihydrogen phosphate

M-Clarity™ quality level = MQ500

3871-10ML
3871-10MLEA 178 EUR
3871-10ML
Naphthol AS-BI phosphate solution 12,5 mg/mL, Sigma-Aldrich®
Naphthol AS-BI phosphate

Naphthol AS-BI Phosphoric acid solution for use in the Sigma Acid Phosphatase (TRAP) Procedure, Kit 387A.


  • Suitable for haematology
  • Suitable for histology

Kaava: C₁₈H₁₅BrNO₆P
MW: 452.1926 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00004058
CAS-numero: 1919-91-1
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR