Tropaeolin O

Toimittaja: Thermo Fisher Scientific

38708.09 38708.04
38708.09EA 122 EUR
38708.09 38708.04
Tropaeolin O
Tropaeolin O

CAS No.: 547-57-9 Molecular Formula: C12H9N2NaO5S Formula Weight: 316.27 Physical Form: Powder Appearance: Orange to brown Solubility: Soluble in water and alcohol MDL No.: MFCD00007499

Kaava: C₁₂H₉N₂O₅SNa
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007499
CAS-numero: 547-57-9
EINECS: 208-924-8
Merck Index: 14,9775

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR