Dishes, evaporating, classy Carbon
Toimittaja: Thermo Fisher Scientific
38372.KT
38372.KTEA 1890 EUR
38372.KT
Dishes, evaporating, classy Carbon
Haihdutusmaljat/Tarjottimet/Altaat Haihdutusaltaat/-maljat
Evaporating dishes, glassy Carbon.
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
Vessels for ultra-high purity materials technology, e.g. semiconductor connections and crystal­growing, e.g. doped halogenide crystals.