Platina 1% on carbon wetted with ca. 50% water, rakeita, reduced

Toimittaja: Thermo Fisher Scientific

38343.14 38343.22
38343.14EA 470 EUR
38343.14 38343.22
Platina 1% on carbon wetted with ca. 50% water, rakeita, reduced
Platina

CAS No.: 7440-06-4 Physical Form: 4x10 Mesh Appearance: Dark gray MDL No.: MFCD00011179

Kaava: Pt
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011179
CAS-numero: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR