Karl Fischer reagents, buffer for bases, HYDRANAL™
Toimittaja: Honeywell Chemicals
37859-1L 37859-6X1L
37859-1LEA 104.01 EUR
37859-1L 37859-6X1L
Karl Fischer reagents, buffer for bases, HYDRANAL™
Karl Fischer-titrantit ja -reagenssit
Liquid buffer medium for Karl Fischer titration in alkaline samples (based on salicylic acid).