Booritribromidi 1 M in heptane, AcroSeal®

Toimittaja: Thermo Fisher Scientific
Vaara

378191000 378198000
378191000.EA 90.6 EUR
378191000. 378198000.
Booritribromidi 1 M in heptane, AcroSeal®
Booritribromidi
Kaava: BBr₃
Kiehumispiste: 90 °C
Tiheys: 0,869 g/cm³
Leimahduspiste: –1 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011312
CAS-numero: 10294-33-4
EINECS: 233-657-9
UN: 3384
ADR: 6.1,

Order Now

Testitulosten erittely

Appearance Clear yellow to amber Solution
Strength 0.9 to 1.1 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR