Sulfuryylikloridi 1.0 M in dichloromethane, AcroSeal®

Toimittaja: Thermo Fisher Scientific
Vaara

378151000 378158000
378151000.EA 70.1 EUR
378151000. 378158000.
Sulfuryylikloridi 1.0 M in dichloromethane, AcroSeal®
Sulfuryylikloridi
Kaava: Cl₂O₂S
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011451
CAS-numero: 7791-25-5
EINECS: 232-245-6
UN: 2922
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Solution
Titration Argentometric 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR