trans-1,2-diaminosykloheksaani-N,N,N',N'-tetraetikkahappomonohydraatii 97.5-100.5% ACS

Toimittaja: Thermo Fisher Scientific
Varoitus

36697.06 36697.14 36697.22 36697.36
36697.06EA 84.5 EUR
36697.06 36697.14 36697.22 36697.36
trans-1,2-diaminosykloheksaani-N,N,N',N'-tetraetikkahappomonohydraatii 97.5-100.5% ACS
trans-1,2-diaminosykloheksaani-N,N,N',N'-tetraetikkahappomonohydraatii

Purity: 97.5%-100.5% CAS No.: 125572-95-4 Molecular Formula: C14H22N2O8.H2O Formula Weight: 364.35 (346.34anhy) Physical Form: Solid Melting Point: 213-216° Beilstein Reference: 3222748 MDL No.: MFCD00066429

Kaava: C₁₄H₂₂N₂O₈·1H₂O
Sulamispiste: 213…216 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00066429
CAS-numero: 125572-95-4
EINECS: 236-308-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR