Iron nickel ≥99.5% (metals basis), jauhe -325 mesh

Toimittaja: Thermo Fisher Scientific
Vaara

36683.22 36683.36 36683.14
36683.22EA 218 EUR
36683.22 36683.36 36683.14
Iron nickel ≥99.5% (metals basis), jauhe -325 mesh
Iron nickel

Purity: 99.5+% Molecular Formula: Ni:Fe; 50:50 wt% Physical Form: Powder Appearance: Grey Odour: Odourless UN No.: UN3089 MDL No.: MFCD01741928


Applications: Low expansion alloy for precision instruments, measuring tapes, weights, etc

Kaava: Ni:Fe;₅₀:₅₀%
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01741928
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR