Aluminum calcium isopropoxide 10% (w/v) in isopropanol (IPA) , 99.8% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

36594.ACEA 213 EUR
36594.AC 36594.AE
Aluminum calcium isopropoxide 10% (w/v) in isopropanol (IPA) , 99.8% (metals basis)
Aluminum calcium isopropoxide

Purity: 99.8% CAS No.: 23275-27-6 Molecular Formula: Ca[Al(OCH(CH3)2)4]2 Formula Weight: 566.30 Physical Form: Liquid Sensitivity: Moisture Sensitive Odour: Alcohol-like Boiling Point: 230-240°/4.0mm UN No.: UN1219 MDL No.: MFCD00143586

Kaava: Ca[Al(OCH(CH₃)₂)₄]₂
Kiehumispiste: 230…240 °C (4,0 mmHg)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00143586
CAS-numero: 23275-27-6
UN: 1219
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR