Jodivetyhappo 47% stabiloitu ACS
Toimittaja: Thermo Fisher Scientific
Vaara
Stabilization: stab. with hypophosphorous acid 1,5%
36490.AE 36490.AP
36490.AEEA 184 EUR
36490.AE 36490.AP
Jodivetyhappo 47% stabiloitu ACS
Jodivetyhappo
CAS No.: 10034-85-2 Molecular Formula: HI Formula Weight: 127.91 Physical Form: Liquid Density: 1.50 Sensitivity: Light Sensitive Appearance: Colourless to yellow Merck Reference: 14,4776 UN No.: 1787 MDL No.: MFCD00011347
Kaava: HI
Kiehumispiste: 127 °C (1013 hPa)
Tiheys: 1,50
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011347
CAS-numero: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8,II
Merck Index: 14,4776
Order Now
TEKNISET TIEDOT
Environmentally Preferable