Supelco® Organic Reference Standard, Loganin

Toimittaja: Merck

36483-10MG-F
36483-10MG-FEA 311 EUR
36483-10MG-F
Supelco® Organic Reference Standard, Loganin
Standardiaineet ja vertailumateriaalit Organic Standards

Pakkaus: Bottomless glass bottle. Contents are inside inserted fused cone.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c3ebb40157b79112d28a7f00c1e3ea83a9e4a11&vid=6c3ebb40157b79112d28a7f00c1e3ea83a9e4a11&prid=web&clid=SAPDAM

Order Now

TEKNISET TIEDOT

Synonym(s) 7-Hydroxy 6-desoxyverbenalin; Loganoside
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nro 18524-94-2
MDL No. MFCD00075645
Kaava C₁₇H₂₆O₁₀
Molekyylipaino 390,38
Leimahduspiste Not applicable
Shipping conditions STD
Säilytysolosuhteet 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR