Atselaiinihappo ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

36308.14 36308.22 36308.59
36308.14EA 115 EUR
36308.14 36308.22 36308.59
Atselaiinihappo ≥98%
Atselaiinihappo

Purity: 98% CAS No.: 123-99-9 Molecular Formula: HOOC(CH2)7COOH Formula Weight: 188.22 Flash Point: 215°(419°F) Physical Form: Crystalline Density: 1.225 Melting Point: ca 102° Boiling Point: 286°/100mm Beilstein Reference: 1101094 Merck Reference: 14,905 MDL No.: MFCD00004432

Kaava: C₉H₁₆O₄
Kiehumispiste: 286 °C (100 mmHg)
Sulamispiste: ∼ 102 °C
Tiheys: 1,225
Leimahduspiste: 215 °C (419 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004432
CAS-numero: 123-99-9
EINECS: 204-669-1
Merck Index: 14,905

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR