Kromi(III)oksidi ≥99.6% (metals basis), fused lump 3-6mm

Toimittaja: Thermo Fisher Scientific
Varoitus

36258.14 36258.22
36258.14EA 129 EUR
36258.14 36258.22
Kromi(III)oksidi ≥99.6% (metals basis), fused lump 3-6mm

For vacuum deposition


It can be used in selective oxidation. It is mainly used for smelting metal chromium and chromium carbide. It is used as coloring agent for color painting on enamelware and porcelain ware, imitation leather or building material. It is also used for making sun-proof coating, abrasive, green polishing compound and special printing ink for printing bank note. It is used as catalyst for organic synthetic chromium.


For vacuum deposition

Kaava: Cr₂O₃
Kiehumispiste: 3000 °C
Sulamispiste: 2435 °C
Tiheys: 5,22
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010949
CAS-numero: 1308-38-9
EINECS: 215-160-9
Merck Index: 14,2234

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR