Aniliini 99+% ACS

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Aminobenzene, Phenylamine

36238.AE 36238.AP 36238.K2
36238.AEEA 76.2 EUR
36238.AE 36238.AP 36238.K2
Aniliini 99+% ACS
Aniliini

Purity: 99+% CAS No.: 62-53-3 Molecular Formula: C6H7N Formula Weight: 93.13 Flash Point: 70°(158°F) Physical Form: Liquid Density: 1.021 Refractive Index: 1.5860 Melting Point: -6° Boiling Point: 183-184° Beilstein Reference: 605631 Merck Reference: 14,659 UN No.: 1547 MDL No.: MFCD00007629

Kaava: C₆H₅NH₂
MW: 93.13 g/mol
Kiehumispiste: 183…184 °C
Sulamispiste: –6 °C
Tiheys: 1.021
Leimahduspiste: 70 °C (158 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007629
CAS-numero: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1 II
Merck Index: 14,659
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR