Natriumkarbonaattimonohydraatii 99.5-100.5% (by anhydrous basis), kiteinen NF, FCC, J.T.Baker®

Toimittaja: Avantor
Varoitus

3600-01EA 97.09 EUR
3600-01 3600-05 3600-09
Natriumkarbonaattimonohydraatii 99.5-100.5% (by anhydrous basis), kiteinen NF, FCC, J.T.Baker®
Natriumkarbonaattimonohydraatii
Kaava: Na₂CO₃·1H₂O
Sulamispiste: 107,85 °C
Tiheys: 2,25 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149177
CAS-numero: 5968-11-6
EINECS: 207-838-8

Order Now

Testitulosten erittely

Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) 12.0 - 15.0 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying 12.0 - 15.0 %
FCC - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR