Zirconium carbide ≥99.5% (metals basis excluding Hf) (max. 200 ppm Hf)

Toimittaja: Thermo Fisher Scientific
Varoitus

35808.18 35808.30
35808.18EA 221 EUR
35808.18 35808.30
Zirconium carbide ≥99.5% (metals basis excluding Hf) (max. 200 ppm Hf)
Zirconium carbide

Purity: 99.5% CAS No.: 12070-14-3 Molecular Formula: ZrC Formula Weight: 103.23 Physical Form: -325 Mesh Powder Density: 6.73 Appearance: Grey Odour: Odourless Melting Point: 3540° UN No.: 3178 MDL No.: MFCD00049640

Kaava: CZr
Sulamispiste: 3540 °C
Tiheys: 6,73
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049640
CAS-numero: 12070-14-3
EINECS: 235-125-1
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR