Samariumtrikloridi, ultra dry ≥99.9% (REO, rare earth oxide basis)

Toimittaja: Thermo Fisher Scientific
Varoitus


Ampuled under argon

35804.04 35804.09 35804.18
35804.04EA 155 EUR
35804.04 35804.09 35804.18
Samariumtrikloridi, ultra dry ≥99.9% (REO, rare earth oxide basis)
Samariumtrikloridi

Purity: 99.9% CAS No.: 10361-82-7 Molecular Formula: SmCl3 Formula Weight: 256.71 Physical Form: Powder, Ampouled under argon Density: 4.46 Melting Point: 678° Boiling Point: dec. Merck Reference: 14,8347 MDL No.: MFCD00011235

Kaava: Cl₃Sm
Kiehumispiste: Dec.
Sulamispiste: 678 °C
Tiheys: 4,46
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011235
CAS-numero: 10361-82-7
EINECS: 233-797-0
Merck Index: 14,8347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR